ContactUsLinkComponent
g-trap ion exchange agarose fast flow columns are ready to use prepacked columns for separation of biomolecules or proteins on the basis of charge. g-trap ion exchange agarose fast flow columns are used for method scouting, separation or purificati 1 * 1 mL
Catalog # GENO786-1010
Supplier:  G-Biosciences
g-trap ion exchange agarose fast flow columns are ready to use prepacked columns for separation of biomolecules or proteins on the basis of charge. g-trap ion exchange agarose fast flow columns are used for method scouting, separation or purificati 1 * 1 mL
Catalog # GENO786-1010
Supplier:  G-Biosciences
Related Information
Frequently Bought Together