ContactUsLinkComponent
the blocking agents trial packs are a set of four g-biosciences proprietary blocking agents for immunoassays with one being a non-an imal protein blocker and another protein free that minimizes the risk of non-specific binding of antibodies during th 1 * 125 mL
Catalog # GENO786-946P
Supplier:  G-Biosciences
the blocking agents trial packs are a set of four g-biosciences proprietary blocking agents for immunoassays with one being a non-an imal protein blocker and another protein free that minimizes the risk of non-specific binding of antibodies during th 1 * 125 mL
Catalog # GENO786-946P
Supplier:  G-Biosciences