Each impact-resistant 50 mL tube contains fifty (50) 1/4 (6.35 mm) diameter Zirconium Oxide coated ceramic grinding spheres. Use to break hard/brittle samples. Specialty applications include RNA extraction from difficult samples where lysis by impac 1 * 500 mL
Catalog # 116959010.
Supplier: MP Biomedicals
Frequently Bought Together