ContactUsLinkComponent
Tubing threaded 75x130x3.2mm gl80 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
  WITG0.182.080
Tubing threaded 75x130x3.2mm gl80 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
  WITG0.182.080
Frequently Bought Together