ContactUsLinkComponent
polystar® 620 DSM-R Bench-type crimp sealer with feed-in at right-hand side, for continuous sealing of sterilization pouches (paper/paper, paper/plastic), casing made of stainless steel, crimp seam width 12 mm, seal speed approx. 6,5 m/min., with ove 1 * 1 items
Catalog # RISC150220
Supplier:  RISCHE HERFURTH
polystar® 620 DSM-R Bench-type crimp sealer with feed-in at right-hand side, for continuous sealing of sterilization pouches (paper/paper, paper/plastic), casing made of stainless steel, crimp seam width 12 mm, seal speed approx. 6,5 m/min., with ove 1 * 1 items
Catalog # RISC150220
Supplier:  RISCHE HERFURTH
Frequently Bought Together