Storage Boxes
5×10, 140×70×36 mm, 12 mm
Catalog # LICEV6-21
Supplier: LICEFA KUNSTSTOFFVERARBEITUNG
Jump to:
Product Details & Documents
Boxes, made of PS, clear, with loose inserts.
Specifications
- For tubes:12 mm
- No. of Holes:5×10
- Dimensions:140×70×36 mm
- Seq:03
- Cat. No.:LICEV6-21