ContactUsLinkComponent
Lens Cleaning Tissues. Non-abrasive cleaning for delicate optical surfaces, lens and optics. Sheets are 100 x 150 mm size. Pack consists of 20 books of 100 sheets. (4 inner packs of 5 books) 1 * 1 PAC
Catalog # HEATHS15920
Supplier:  HEATHROW
undefined
Lens Cleaning Tissues. Non-abrasive cleaning for delicate optical surfaces, lens and optics. Sheets are 100 x 150 mm size. Pack consists of 20 books of 100 sheets. (4 inner packs of 5 books) 1 * 1 PAC
Catalog # HEATHS15920
Supplier:  HEATHROW
Frequently Bought Together