Orders LinkContactUsLinkComponent
Personal padlock kit
Personal padlock kit
Personal padlock kit, incl. 1× Safety lockout hasp, 1× key holder, 2× safety padlocks
Catalog # BRDY873872
Supplier:   Brady
Personal padlock kit
Catalog # BRDY873872
Supplier:  Brady
Personal padlock kit
Personal padlock kit
Personal padlock kit, incl. 1× Safety lockout hasp, 1× key holder, 2× safety padlocks
Price
109,00 €
Log in for availability
1 * 1 items
Product Details & Documents
This padlock kit includes the equipment needed to implement lockout safety procedures. A convenient way to carry padlocks, hasps and keys in a handy pouch.
  • Pouch has a clear face allowing for easy identification
  • Heavy duty retractable key holder features a Kevlar® cord with split ring attachment and a convertible belt clip/belt loop back attachment
  • Key holder is extendable to 1200 mm
  • Clip can be attached to pouch or be used independently
Delivery information: The kit includes: One Safety lockout hasp (25 mm shackle width), one Heavy Duty retractable key holder and two red safety padlocks.
Specifications
  • Type:
    Personal padlock kit, incl. 1× Safety lockout hasp, 1× key holder, 2× safety padlocks
  • Seq:
    01