Padlock stations
Small padlock station, for 4 padlocks, Red, 76×31×57 mm
Catalog # BRDY800120
Supplier: Brady
Jump to:
Product Details & Documents
These stations constructed of 14 gauge steel with red plastic coating are ideal for storing padlocks when they are not in use.
- Rugged and eye-catching stations
- Hold up to 4 or 24 padlocks
Ordering information: Padlocks supplied seperately.
Specifications
- Colour:Red
- Description:Small padlock station, for 4 padlocks
- Seq:01
- W×D×H:76×31×57 mm