Push button lockout devices
Small (for 16 mm push buttons), Clear
Catalog # BRDY139793
Supplier: Brady
Jump to:
Product Details & Documents
These lockout devices prevent the unauthorised use of push button emergency devices.
- With standard cover
- Made from clear PC, with translucent red cover
- Suitable for 16, 22 or 30 mm push buttons
Specifications
- Type:Small (for 16 mm push buttons)
- Colour:Clear
- Seq:02