Fuse blockout devices
Fuse blockout device, large, for 20,6 mm, 27 mm, and blade type fuses, Red
Catalog # BRDY065691
Supplier: Brady
Jump to:
Product Details & Documents
These highly visible fuse blockout devices made from PP can be used in conjunction with Brady lockout tags.
Documents
Specifications
- Type:Fuse blockout device, large, for 20,6 mm, 27 mm, and blade type fuses
- Colour:Red
- Seq:02