Fuse blockout devices
Fuse blockout device, small, for 6,3 / 7,1 / 10,3 / 14,3 mm fuses, Red
BRDY065690
: Brady
These highly visible fuse blockout devices made from PP can be used in conjunction with Brady lockout tags.
- Type:Fuse blockout device, small, for 6,3 / 7,1 / 10,3 / 14,3 mm fuses
- Colour:Red
- Seq:01