Beaker Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), 250 ml volume, wide form, with lip and spout, tr anslucent, high chemical resistance and purity. Diameter 68mm, height 88mm. The durability or operating cycles of our q 1 * 1 items
Catalog # TSLA73161
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together