Crucible Quartz glass for coking in application temperatures of up to 1050°C (maximum 1600°C), without handle, for volatille matter , translucent, high chemical resistance and purity. Diameter 25mm, height 38mm. The durability or operating cycles of 1 * 1 items
Catalog # TSLA73088
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together