ContactUsLinkComponent
Capsule Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), circular, low form, with lid, translucent, hig h chemical resistance and purity. Diameter 75mm, height 12mm. The durability or operating cycles of our Capsule Quartz g 1 * 1 items
Catalog # TSLA73033
Capsule Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), circular, low form, with lid, translucent, hig h chemical resistance and purity. Diameter 75mm, height 12mm. The durability or operating cycles of our Capsule Quartz g 1 * 1 items
Catalog # TSLA73033
Frequently Bought Together