ContactUsLinkComponent
Capsule-tobacco ashin Quartz glassg in application temperatures of up to 1050°C (maximum 1600°C), translucent, high chemical resis tance and purity. Length 78mm, width 56mm, height 285mm. The durability or operating cycles of our Capsule Quartz glass 1 * 1 items
Catalog # TSLA73027
Capsule-tobacco ashin Quartz glassg in application temperatures of up to 1050°C (maximum 1600°C), translucent, high chemical resis tance and purity. Length 78mm, width 56mm, height 285mm. The durability or operating cycles of our Capsule Quartz glass 1 * 1 items
Catalog # TSLA73027
Frequently Bought Together