ContactUsLinkComponent
The PTFE Template tool allows a fixed area to be swabbed. A PTFE template designed to a predetermined area ensures swabbing is carried out over a set area. This gives improved results. Hole 5x5 cm. 1 * 1 items
Catalog # 300-0418
Supplier:  Sampling Systems
The PTFE Template tool allows a fixed area to be swabbed. A PTFE template designed to a predetermined area ensures swabbing is carried out over a set area. This gives improved results. Hole 5x5 cm. 1 * 1 items
Catalog # 300-0418
Supplier:  Sampling Systems
Frequently Bought Together