Covernets
Replacement net for 215×206 mm
Catalog # MIEL10283690
Supplier: MIELE
Jump to:
Product Details & Documents
Covering net for optimum fixing of delicate items to be washed.
- Stainless steel frame covered with plastic net
- Particularly resistant and durable
Specifications
- Description:Replacement net for 215×206 mm
- Seq:04