testo 350 analyzer box, eqipped with O2, incl. differential pressure sensor, temperature probe input Type K NiCr-Ni and Type S Pt10Rh-Pt, connection Testo databus, rech. battery, integrated combustion air probe (NTC), trigger input, measurement data 1 * 1 items
Catalog # TEST0632.3510
Supplier: Testo
Frequently Bought Together