CF™583 succinimidyl ester is a superior deep-red fluorescent dye with an amine-reactive group. CF™583 is spectrally identical to Cy® 3.5 but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. 1 * 1 µmol
Catalog # 96016.
Supplier: Biotium
Frequently Bought Together