ContactUsLinkComponent
SONOREX SUPER ZE 1031 DT, ultrasonic built-in unit with ultrasound from the bottom, 35 kHz, 230 V( 10%) 50/60 Hz, consisting of s/s oscillating tank with bead 1 1/2 , HF-generator and timer 1-30 min and continuous and temperature control ID 510x30 1 * 1 items
Catalog # BAND3217-UK
SONOREX SUPER ZE 1031 DT, ultrasonic built-in unit with ultrasound from the bottom, 35 kHz, 230 V( 10%) 50/60 Hz, consisting of s/s oscillating tank with bead 1 1/2 , HF-generator and timer 1-30 min and continuous and temperature control ID 510x30 1 * 1 items
Catalog # BAND3217-UK
Frequently Bought Together