ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed goat anti-chicken IgY (H+L) with horseradish peroxidase. To minimize cros s-reactivity, the antibody has been adsorbed against bovine, goat, guinea pig, Syrian hamster, horse, human, mouse, r 1 * 1 mL
Catalog # 20474-1ML
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed goat anti-chicken IgY (H+L) with horseradish peroxidase. To minimize cros s-reactivity, the antibody has been adsorbed against bovine, goat, guinea pig, Syrian hamster, horse, human, mouse, r 1 * 1 mL
Catalog # 20474-1ML
Supplier:  Biotium
Frequently Bought Together