ContactUsLinkComponent
GasAGasAlert MicroClip XL 2-Gas H2S / CO 2-gas-detector diffusion device Sensors: CO: electrochemical Sensor, Measuring range : 0-500ppm H2S: electrochemical Sensor, Measuring range: 0-100ppm Additional GasAlertMicroClip features: • Choose the GasAle 1 * 1 items
Catalog # BWTEMCXL-00HM-Y-EU
Supplier:  BW TECHNOLOGIES
GasAGasAlert MicroClip XL 2-Gas H2S / CO 2-gas-detector diffusion device Sensors: CO: electrochemical Sensor, Measuring range : 0-500ppm H2S: electrochemical Sensor, Measuring range: 0-100ppm Additional GasAlertMicroClip features: • Choose the GasAle 1 * 1 items
Catalog # BWTEMCXL-00HM-Y-EU
Supplier:  BW TECHNOLOGIES
Frequently Bought Together