Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
Slide box for micro slides, Black/cream, Wood/cardboard, 100
Catalog # HECH42701010
Slide boxes
Catalog # HECH42701010
Slide boxes
Slide boxes
Slide box for micro slides, Black/cream, Wood/cardboard, 100
Price
18,50 €
Log in for availability
1 * 1 items
Product Details & Documents
These slide boxes are designed for secure, dust-free storage of specimen slides with metal catch and hinges.
  • Numbered index in the lid
  • Cardboard with black wooden frame
  • Numbered plastic grooves: Anti-static and xylene-resistant
Suitable for the safe and protected storage of slides.
Documents
Specifications
  • No. of slides:
    100
  • Colour:
    Black/cream
  • Description:
    Slide box for micro slides
  • Material:
    Wood/cardboard
  • Seq:
    05