ContactUsLinkComponent
HIGH VOLUME, FAST-LOADING PFA FAST LOOPS FOR HIGH-FLOW VALVES. RECOMMENDED FOR USE WITH 1.0 MM I.D. (GRAY) PROBES AND APPLICATIONS R EQUIRING LARGER SAMPLE VOLUMES OR HIGH FLOW RATES. 3.2 MM O.D., 1/4-28 FITTINGS. 1 * 1 items
Catalog # PERCN0777653
Supplier:  PerkinElmer
HIGH VOLUME, FAST-LOADING PFA FAST LOOPS FOR HIGH-FLOW VALVES. RECOMMENDED FOR USE WITH 1.0 MM I.D. (GRAY) PROBES AND APPLICATIONS R EQUIRING LARGER SAMPLE VOLUMES OR HIGH FLOW RATES. 3.2 MM O.D., 1/4-28 FITTINGS. 1 * 1 items
Catalog # PERCN0777653
Supplier:  PerkinElmer
Frequently Bought Together