Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEINHARD SILQ PLUS ULTRA HIGH PURITY QUARTZ NEBULIZER, LOW INTERNAL VOLUME, 0.5 ML/MIN, 50 PSI, WITH FG+ PFA GAS FITTING AND SAMPLE UPTAKE LINE. 1 * 1 items
Catalog # PERCN8122460
Supplier:
PerkinElmer
Expand image
MEINHARD SILQ PLUS ULTRA HIGH PURITY QUARTZ NEBULIZER, LOW INTERNAL VOLUME, 0.5 ML/MIN, 50 PSI, WITH FG+ PFA GAS FITTING AND SAMPLE UPTAKE LINE. 1 * 1 items
Catalog # PERCN8122460
Supplier:
PerkinElmer
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection