ContactUsLinkComponent
ATEX CATEGORY 2 ZONES 1-2 (GAS) & ZONES 21-22 (DUST) MAXIMUM SURFACE TEMPERATURE : T6, 85°C AMBIANT TEMPERATURE : +40°C / -20°C Air flow: 187 m3/h Vacuum pressure: 4572 mmH2O Recovery Capacity: 23L - Dry recovery. 28L – Wet recover Dimensions 1 * 1 items
Catalog # TIGV110366
Supplier:  TIGER-VAC
ATEX CATEGORY 2 ZONES 1-2 (GAS) & ZONES 21-22 (DUST) MAXIMUM SURFACE TEMPERATURE : T6, 85°C AMBIANT TEMPERATURE : +40°C / -20°C Air flow: 187 m3/h Vacuum pressure: 4572 mmH2O Recovery Capacity: 23L - Dry recovery. 28L – Wet recover Dimensions 1 * 1 items
Catalog # TIGV110366
Supplier:  TIGER-VAC
Frequently Bought Together