Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Goat anti-Mouse IgG Fc, TRITC conjugated, min. cross-reactivity to bovine, horse or human serum proteins 1 * 1 mg
Catalog # AGRIAS163484
Supplier:
Agrisera
Expand image
Goat anti-Mouse IgG Fc, TRITC conjugated, min. cross-reactivity to bovine, horse or human serum proteins 1 * 1 mg
Catalog # AGRIAS163484
Supplier:
Agrisera
Related Information
Documentation
About Agrisera
Documentation
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals