Jalas Clogs Occupational 5012 Menu White, Regular Fit, Microfiber, Hygienical Liner, Low Weight, oil resistant outsole, anti-static properties, adjustable heel strap, double shock absorp zones, ESD, washable 40degC, Indoor, Slippery or dry environmen 1 * 1 PAIR
EJEN5012-45
: EJENDALS
Frequently Bought Together