Orders LinkContactUsLinkComponent
Cut resistant gloves, A620
Cut resistant gloves, A620
9, Grey
Catalog # PRTWA620GRRL
Supplier:   Portwest
Cut resistant gloves, A620
Catalog # PRTWA620GRRL
Supplier:  Portwest
Cut resistant gloves, A620
Cut resistant gloves, A620
9, Grey
Price
61,60 €
Log in for availability
1 * 12 PAIR
Product Details & Documents
These seamless HPPE/glass fibre gloves with a palm PU coating provide dexterity, cut resistance and good grip.
  • Smooth PU coating for increased abrasion resistance
  • Palm dipped to increase dexterity and ventilation
  • 100% breathable seamless liner
  • 13 gauge liner for a perfect fit
  • Low linting construction for minimal contamination
Certifications: EN 388:2016 (4241BX), EN 407 (X1XXX).
Specifications
  • Glove size:
    9
  • Colour:
    Grey
  • Seq:
    04
  • Cat. No.:
    PRTWA620GRRL
  • Liner material:
    HPPE
  • Disposable / reusable:
    Reusable
  • Model:
    A620
  • Cuff style:
    Knit wrist
  • EN norm:
    EN 388:2016 (4241BX), EN 407 (X1XXX)
  • Finish:
    ¾ coating
  • Ambidextrous / hand-specific:
    Hand-specific
  • Gauge:
    13
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.