ContactUsLinkComponent
Biological Indicatorological Indicator Spore Strip for Monitoring Chlorine Dioxide Processes, Geobacillus stearothermophilus Cell Li ne 7953, Each 6 mm x 30 mm Strip Individually Packaged in Tyvek®/Tyvek® Pouches, 100 per box, 106 population level, I 1 * 1 Each
Catalog # NAMSTCDS-06
Supplier:  CROSSTEX
Biological Indicatorological Indicator Spore Strip for Monitoring Chlorine Dioxide Processes, Geobacillus stearothermophilus Cell Li ne 7953, Each 6 mm x 30 mm Strip Individually Packaged in Tyvek®/Tyvek® Pouches, 100 per box, 106 population level, I 1 * 1 Each
Catalog # NAMSTCDS-06
Supplier:  CROSSTEX
Frequently Bought Together