ContactUsLinkComponent
Steam Formaldehyde Biological Indicatorological Indicator strip with , Geobacillus stearothermophilus Cell Line 7953. 6 mm x 30 mm Strips Individually Packaged in Glassine Pouches, 100 strips per box, 106 population level, ISO 11138-1, 11138-3 and US 1 * 1 Each
  NAMSSTSF-06
 :  CROSSTEX
Steam Formaldehyde Biological Indicatorological Indicator strip with , Geobacillus stearothermophilus Cell Line 7953. 6 mm x 30 mm Strips Individually Packaged in Glassine Pouches, 100 strips per box, 106 population level, ISO 11138-1, 11138-3 and US 1 * 1 Each
  NAMSSTSF-06
 :  CROSSTEX
Frequently Bought Together