ContactUsLinkComponent
Biological Indicatorological Indicator spore discs for monitoring hydrogen peroxide/plasma processes, Bacillus atrophaeus Cell Line 9372, 9 mm glass fiber discs individually packaged in Tyvek/Mylar envelopes, 100 per box, 105 population level, ISO 11 1 * 1 Each
Catalog # NAMSGFTN-05
Supplier:  CROSSTEX
Biological Indicatorological Indicator spore discs for monitoring hydrogen peroxide/plasma processes, Bacillus atrophaeus Cell Line 9372, 9 mm glass fiber discs individually packaged in Tyvek/Mylar envelopes, 100 per box, 105 population level, ISO 11 1 * 1 Each
Catalog # NAMSGFTN-05
Supplier:  CROSSTEX
Frequently Bought Together