ContactUsLinkComponent
Spore Thread Biological Indicatorological Indicators for Monitoring Ethylene Oxide (EO) or Dry Heat Processes, Bacillus atrophaeus C ell Line 9372, Cotton Threads each 25mm in length, Individually Packaged in Glassine Envelopes, 100 per box, 106 pop 1 * 1 Each
Catalog # NAMSTHN-06P
Supplier:  CROSSTEX
Spore Thread Biological Indicatorological Indicators for Monitoring Ethylene Oxide (EO) or Dry Heat Processes, Bacillus atrophaeus C ell Line 9372, Cotton Threads each 25mm in length, Individually Packaged in Glassine Envelopes, 100 per box, 106 pop 1 * 1 Each
Catalog # NAMSTHN-06P
Supplier:  CROSSTEX
Frequently Bought Together