ContactUsLinkComponent
Digital Muffle Furnace, programmable, 1000C, 14 Liter, with 4-side heating, digital PID Control System LCD, Timer: 99 hr. / 59 min., internal dim.: 250 x 350 x 160 mm, external dim.: 530 x 580 x 620 mm, power supply 400 V, 50/60 Hz, power plug 3 ph 1 * 1 items
Catalog # WITGDH.WF23.P14
Supplier:  witeg Labortechnik
Digital Muffle Furnace, programmable, 1000C, 14 Liter, with 4-side heating, digital PID Control System LCD, Timer: 99 hr. / 59 min., internal dim.: 250 x 350 x 160 mm, external dim.: 530 x 580 x 620 mm, power supply 400 V, 50/60 Hz, power plug 3 ph 1 * 1 items
Catalog # WITGDH.WF23.P14
Supplier:  witeg Labortechnik
Frequently Bought Together