ISOLATION GOWN COLOR: GREEN SIZE: 120X140 DESCRIPTION: WITH TIES ON NECK AND WRIST MATERIAL: POLYPROPYLENE THICKNESS: 17 G/M PACKAGING: 5 PCS/BAG, 100 PCS/CARTON 1 * 100 items
Catalog # GUNA140144
Supplier: VWR Collection
Frequently Bought Together