Dilatometers, Quickfit®
Dilatometer stopper with cone size NS 14/23
Catalog # BIBBSB14DL
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Dilatometer body is graduated with an arbitrary scale
- Interchangable stopper has open top to allow weighting with steel balls etc.
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place.
: The body and stopper should be ordered separately.
Specifications
- Description:Dilatometer stopper with cone size NS 14/23
- Seq:02