Quartz inceneration crucible per Rademacher in application temperatures of up to 1050°C, transparent, high chemical resistance and purity. Diameter 27mm, height 45mm. The durability or operating cycles of our quartz glass product mainly depends on ap 1 * 1 items
Catalog # TSLA75702
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together