ContactUsLinkComponent
Quartz inceneration crucible in application temperatures of up to 1050°C, transparent, high chemical resistance and purity. Diamete r 25mm, height 38mm. The durability or operating cycles of our quartz glass product mainly depends on applications. Fo 1 * 1 items
Catalog # TSLA75700
Quartz inceneration crucible in application temperatures of up to 1050°C, transparent, high chemical resistance and purity. Diamete r 25mm, height 38mm. The durability or operating cycles of our quartz glass product mainly depends on applications. Fo 1 * 1 items
Catalog # TSLA75700
Frequently Bought Together