Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RACK 4-WAY INTERLOCKING COMBIRACK ASSORTED COLOURS AUTOCLAVABLE PACK OF 5 WITH 4 ORIENTATIONS TO ACCOMMODATE 0.5ml 1.5ml 2.0ml 15ml and 50ml 1 * 5 items
Catalog # ALPCLW5840AS
Supplier:
Alpha Laboratories
Expand image
RACK 4-WAY INTERLOCKING COMBIRACK ASSORTED COLOURS AUTOCLAVABLE PACK OF 5 WITH 4 ORIENTATIONS TO ACCOMMODATE 0.5ml 1.5ml 2.0ml 15ml and 50ml 1 * 5 items
Catalog # ALPCLW5840AS
Supplier:
Alpha Laboratories
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection