ContactUsLinkComponent
RACK 100-POSITION FREEZER RACK HINGED LID 10 X 10 ARRAY ASSORTED TEMPERATURE 90C TO 120C PACK OF 5 FITS 1.5ML 2.0ML MICROTUBES OR 1.2ML CRYOVIALS 1 * 1 items
Catalog # ALPCLW5750AS
Supplier:  Alpha Laboratories
RACK 100-POSITION FREEZER RACK HINGED LID 10 X 10 ARRAY ASSORTED TEMPERATURE 90C TO 120C PACK OF 5 FITS 1.5ML 2.0ML MICROTUBES OR 1.2ML CRYOVIALS 1 * 1 items
Catalog # ALPCLW5750AS
Supplier:  Alpha Laboratories
Frequently Bought Together