ContactUsLinkComponent
Pasteurpipettes, made of neutral soda glass, with the tip closed and cotton in the mouth. Easy opening of the closed part by pushing it slightly. Dimensions: mouth: 7.00 mm tip: 1.4 to 1.6 mm thickness: 0.50 mm 1 * 1.000 items
Catalog # DELT712
Supplier:  DELTALAB
Pasteurpipettes, made of neutral soda glass, with the tip closed and cotton in the mouth. Easy opening of the closed part by pushing it slightly. Dimensions: mouth: 7.00 mm tip: 1.4 to 1.6 mm thickness: 0.50 mm 1 * 1.000 items
Catalog # DELT712
Supplier:  DELTALAB
Frequently Bought Together