CF™440 maleimide is a bluish green fluorescent dye with a maleimide reactive group, which can be used to label a target molecule wi th a thiol group. CF™440 (abs/em=440 nm/515 nm) is a superior alternative to Alexa Fluor 430 owing to its exceptiona 1 * 1 µmol
Catalog # 92124.
Supplier: Biotium
Frequently Bought Together