ContactUsLinkComponent
CF™440 succinimidyl ester (CF™440 SE) is a green fluorescent dye with an amine-reactive group (i.e., succinimidyl ester group). CF™ 440 (abs/em=440 nm/515 nm) is a superior alternative to Alexa Fluor 430 owing to its exceptional photostability. 1 * 1 µmol
  92123.
 :  Biotium
CF™440 succinimidyl ester (CF™440 SE) is a green fluorescent dye with an amine-reactive group (i.e., succinimidyl ester group). CF™ 440 (abs/em=440 nm/515 nm) is a superior alternative to Alexa Fluor 430 owing to its exceptional photostability. 1 * 1 µmol
  92123.
 :  Biotium
Frequently Bought Together