ALKALINE PHOSPHATASE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO HUMAN IgG Fc. Anti-Human Host: goat Form: affinity purified liq uid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. AP Co 1 * 1 mL
Catalog # ICNA0859289
Supplier: MP Biomedicals
Frequently Bought Together