ContactUsLinkComponent
ANISOMYCIN. From Streptomyces griseolusPurity: 97%Activates a restricted subset of kinases currently only identified by p54 and MAP kinase. Study aid for cytoplasmic signals resulting in nuclear signaling and induction of c-fos and c-jun .Ref.: 1. Ka 1 * 10 mg
Catalog # ICNA0215988110
Supplier:  MP Biomedicals
ANISOMYCIN. From Streptomyces griseolusPurity: 97%Activates a restricted subset of kinases currently only identified by p54 and MAP kinase. Study aid for cytoplasmic signals resulting in nuclear signaling and induction of c-fos and c-jun .Ref.: 1. Ka 1 * 10 mg
Catalog # ICNA0215988110
Supplier:  MP Biomedicals
Frequently Bought Together