From Chicken Egg white Type VI. Molecular Biology Reagent3X Crystallized Salt-Free, Albumin-Free Lyophilized. Activity: =2 3,500 Shugar units/mgUnit Definition: One unit will produce a decrease in A450 of 0.001 per minute at pH 6.24 and 25C using Mi 1 * 1 g
Catalog # ICNA0219530301
Supplier: MP Biomedicals
Frequently Bought Together