ContactUsLinkComponent
From Canavalia ensiformis Highly Purifed Esssentially Salt Free Possesses specificity for terminal alfa-D -mannosyl and alfa-D -glucosyl residues and also agglutinates red blood cells. Caution: Has mitogenic properties. 1 * 250 mg
Catalog # ICNA02195283.2
Supplier:  MP Biomedicals
From Canavalia ensiformis Highly Purifed Esssentially Salt Free Possesses specificity for terminal alfa-D -mannosyl and alfa-D -glucosyl residues and also agglutinates red blood cells. Caution: Has mitogenic properties. 1 * 250 mg
Catalog # ICNA02195283.2
Supplier:  MP Biomedicals