Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SC 150-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S7, Amb+13 TO +150 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB UL/CSA, 1 * 1 items
Catalog # HAKA1531071
Supplier:
THERMO ELECTRON LED
Expand image
SC 150-S7 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S7, Amb+13 TO +150 C, 4-7 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB UL/CSA, 1 * 1 items
Catalog # HAKA1531071
Supplier:
THERMO ELECTRON LED
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals