Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
SC 150-S5P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC150-S5P, Amb+13 to 100 C, 3-5 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB 1 * 1 items
HAKA1533051
:
THERMO ELECTRON LED
SC 150-S5P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC150-S5P, Amb+13 to 100 C, 3-5 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB 1 * 1 items
HAKA1533051
:
THERMO ELECTRON LED
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals